missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ivabradine hydrochloride

Product Code. 17261212 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17261212 250 mg 250mg
17251212 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17261212 Supplier Thermo Scientific Chemicals Supplier No. 466282500
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 148849-67-6 | C27H36N2O5·HCl, C27H37ClN2O5 | 505.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 148849-67-6
Molecular Formula C27H36N2O5·HCl

Specifications

Quantity 250 mg
Solubility Solubility aq. soln.: .. Other solubilities: soluble in dmso and methanol
Melting Point 140.0°C
Formula Weight 505.05
Chemical Name or Material Ivabradine hydrochloride
compliance-icons
Product Identifier
  • Ivabradine hydrochloride
Signal Word
  • Warning
Hazard Category
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Ivabradine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.