missing translation for 'onlineSavingsMsg'
Learn More

Ivabradine-d3 Hydrochloride, TRC

Product Code. 30403756
Change view
Click to view available options
Quantity:
1 mg
10 mg
Product Code. Quantity
30403756 10 mg
30403216 1 mg
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30403756

Brand: Toronto Research Chemicals TRCI94050210MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1217809-61-4
Formula Weight 507.2579
InChI Formula InChI=1S/C27H36N2O5.ClH/c1-28(17-21-11-20-14-25(33-4)26(34-5)16-22(20)21)8-6-9-29-10-7-18-12-23(31-2)24(32-3)13-19(18)15-27(29)30;/h12-14,16,21H,6-11,15,17H2,1-5H3;1H/t21-;/m1./s1/i1D3;
IUPAC Name 3-[3-[[(7S)-3,4-dimethoxy-7-bicyclo[4.2.0]octa-1,3,5-trienyl]methyl-(trideuteriomethyl)amino]propyl]-7,8-dimethoxy-2,5-dihydro-1H-3-benzazepin-4-one;hydrochloride
Molecular Formula C27 2H3 H33 N2 O5 . Cl H
Molecular Weight (g/mol) 508.06
SMILES Cl.[2H]C([2H])([2H])N(CCCN1CCc2cc(OC)c(OC)cc2CC1=O)C[C@H]3Cc4cc(OC)c(OC)cc34
Synonym 3-[3-[[(7S)-3,4-Dimethoxy-7-bicyclo[4.2.0]octa-1(6),2,4-trienyl]methyl-(trideuteriomethyl)amino]propyl]-7,8-dimethoxy-2,5-dihydro-1H-3-benzazepin-4-one hydrochloride, Ivabradine-d3 Hydrochloride, Ivabradine-D3 Hydrochloride (N-methyl D3)
Chemical Name or Material Ivabradine hydrochloride N-methyl-d3

Specifications

Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.