missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IUPAC pH Standard pH 6.865, Hach Lange™
Click to view available options
Quantity:
500 mL
Unit Size:
Each
Description
- pH Standard, IUPAC, pH10.012 at 25 °C, Certified, 500 mL (Radiometer Analytical)
- 10.012 ± 0.010 pH at 25°C concentration
- Traceable to IUPAC pH scale and NIST
- Shelf life of 48 months from production date
- Each bottle (500 mL) is supplied with a COFRAC Verification Certificate and Certificate of Conformity and Traceability drawn up according to ISO31
- Direct traceability to primary standards and the reference method
- The competence and integrity of a laboratory internationally recognized to be accredited for the measurement of pH and conductivity reference materials
Specifications
Specifications
| pH | 6.865 |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Traceability to NIST | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction