missing translation for 'onlineSavingsMsg'
Learn More

ITX3, MedChemExpress

Product Code. 16489647
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16489647 5 mg 5mg
16467108 10 mg 10mg
16453147 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16489647 Supplier Medchem Express Supplier No. HY16663
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress ITX3 is a specific and nontoxic inhibitor of the TrioN (N-terminal GEF domain of the multidomain Trio protein) with IC50 of 76 uM; inhibits TrioN-stimulated RhoG exchange in vitro.

ITX3 is a specific and nontoxic inhibitor of the TrioN (N-terminal GEF domain of the multidomain Trio protein) with IC50 of 76 uM

Specifications

Chemical Name or Material ITX3
CAS 347323-96-0
Color Light Green
Purity Grade Notes Research
Molecular Formula C22H17N3OS
Quantity 5 mg
Solubility Information DMSO : 2 mg/mL (5.38 mM; ultrasonic and warming and heat to 80°C)
SMILES O=C(/C1=C\C2=C(C)N(C(C)=C2)C3=CC=CC=C3)N4C(S1)=NC5=CC=CC=C45
Molecular Weight (g/mol) 371.45
Formula Weight 371.45
Percent Purity 98.01%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.