missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ ITS-Plus Media Supplement (100X)

Product Code. 18768706
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
Product Code. Quantity unitSize
18768706 5 mL 5mL
1 options
This item is not returnable. View return policy

Product Code. 18768706

Brand: R&D Systems™ AR014

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Designed to support cell proliferation under low serum conditions

Insulin-Transferrin-Selenium-Plus Media Supplement
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Supports cell proliferation under low serum conditions
Quantity 5 mL
Content And Storage Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.