missing translation for 'onlineSavingsMsg'
Learn More

Itraconazole, MedChemExpress

Product Code. 16407356
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16407356 100 mg 100mg
16461876 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16407356 Supplier Medchem Express Supplier No. HY17514100mg
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Itraconazole (R51211) is a triazole antifungal agent and a potent and orally active Hedgehog (Hh) signaling pathway antagonist with an IC50 of ∼800 nM. Itraconazole potently inhibits lanosterol 14α-demethylase (cytochrome P450 enzyme), thereby inhibits the oxidative conversion of lanosterol to ergosterol. Itraconazole has anticancer and antiangiogenic effects. Itraconazole is a oxysterol-binding protein (OSBP) inhibitor.

Itraconazole (R51211) is a triazole antifungal agent and a potent and orally active Hedgehog (Hh) signaling pathway antagonist with an IC50 of ∼800 nM. Itraconazole potently inhibits lanosterol 14α-demethylase (cytochrome P450 enzyme), thereby inhibits the oxidative conversion of lanosterol to ergosterol. Itraconazole has anticancer and antiangiogenic effects. Itraconazole is a oxysterol-binding protein (OSBP) inhibitor.

Specifications

Chemical Name or Material Itraconazole
Physical Form Solid
CAS 84625-61-6
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C35H38Cl2N8O4
For Use With (Application) COVID-19-immunoregulation
Synonym R51211
Solubility Information DMSO : 6.25 mg/mL (8.86 mM; Need ultrasonic)
SMILES ClC1=C(C=CC(Cl)=C1)[C@@]2(O[C@@H](COC3=CC=C(N4CCN(C5=CC=C(N(C=NN6C(C)CC)C6=O)C=C5)CC4)C=C3)CO2)CN7C=NC=N7
Molecular Weight (g/mol) 705.63
Formula Weight 705.63
Percent Purity 99.15%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.