missing translation for 'onlineSavingsMsg'
Learn More

Itraconazole Desethylene-seco-piperazine Di-N-formyl Impurity, TRC

Product Code. 30395195
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395195 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30395195 Supplier Toronto Research Chemicals Supplier No. TRCI9375651MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Itraconazole Desethylene-seco-Piperazine di-N-Formyl
CAS 1199350-02-1
Molecular Formula C35 H36 Cl2 N8 O6
Quantity 1 mg
Synonym rel-N-[2-[[4-[[(2R,4S)-2-(2,4-Dichlorophenyl)-2-(1H-1,2,4-triazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]formylamino]ethyl]-N-[4-[1,5-dihydro-1-(1-methylpropyl)-5-oxo-4H-1,2,4-triazol-4-yl]phenyl]formamide
InChI Formula InChI=1S/C35H36Cl2N8O6/c1-3-25(2)45-34(48)44(22-40-45)29-7-5-27(6-8-29)41(23-46)14-15-42(24-47)28-9-11-30(12-10-28)49-17-31-18-50-35(51-31,19-43-21-38-20-39-43)32-13-4-26(36)16-33(32)37/h4-13,16,20-25,31H,3,14-15,17-19H2,1-2H3/t25?,31-,35-/m0/s1
SMILES CCC(C)N1N=CN(C1=O)c2ccc(cc2)N(CCN(C=O)c3ccc(OC[C@H]4CO[C@@](Cn5cncn5)(O4)c6ccc(Cl)cc6Cl)cc3)C=O
IUPAC Name N-[4-(1-butan-2-yl-5-oxo-1,2,4-triazol-4-yl)phenyl]-N-[2-[4-[[(2R,4S)-2-(2,4-dichlorophenyl)-2-(1,2,4-triazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]-N-formylanilino]ethyl]formamide
Molecular Weight (g/mol) 735.62
Formula Weight 734.2135
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.