Learn More
Itraconazole, 98%, Thermo Scientific™
£113.00 - £527.00
Chemical Identifiers
| CAS | 84625-61-6 |
|---|---|
| Molecular Formula | C35H38Cl2N8O4 |
| Molecular Weight (g/mol) | 705.64 |
| MDL Number | MFCD00870168,MFCD00941396 |
| InChI Key | VHVPQPYKVGDNFY-ZPGVKDDISA-N |
| Synonym | Oriconazole; R51211 |
| PubChem CID | 45039617 |
| IUPAC Name | 1-(butan-2-yl)-4-{4-[4-(4-{[(2R,4S)-2-(2,4-dichlorophenyl)-2-[(1H-1,2,4-triazol-1-yl)methyl]-1,3-dioxolan-4-yl]methoxy}phenyl)piperazin-1-yl]phenyl}-4,5-dihydro-1H-1,2,4-triazol-5-one |
| SMILES | CCC(C)N1N=CN(C1=O)C1=CC=C(C=C1)N1CCN(CC1)C1=CC=C(OC[C@H]2CO[C@@](CN3C=NC=N3)(O2)C2=CC=C(Cl)C=C2Cl)C=C1 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15497729
|
Thermo Scientific Alfa Aesar
J66390.06 |
5g |
£113.00
5g |
Estimated Shipment: 03-09-2026
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
15407739
|
Thermo Scientific Alfa Aesar
J66390.14 |
25g |
£527.00
25g |
Estimated Shipment: 01-10-2026
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsItraconazole is a triazole antifungal agent. It is used to inhibit cytochrome P-450-dependent enzymes and ergosterol synthesis. It has been used against histoplasmosis, blastomycosis, cryptococcal meningitis, and aspergillosis. It′s different formulations are used to study Candida strains in murine invasive infections. It has been used to study proliferative changes of the forestomach mucosa in alloxan-induced diabetic rats..
Solubility
Insoluble in water. Solube in chloroform at 50 mg/ml. Slightly soluble in ethanol or methanol
Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
Chemical Identifiers
| 84625-61-6 | |
| 705.64 | |
| VHVPQPYKVGDNFY-ZPGVKDDISA-N | |
| 45039617 | |
| CCC(C)N1N=CN(C1=O)C1=CC=C(C=C1)N1CCN(CC1)C1=CC=C(OC[C@H]2CO[C@@](CN3C=NC=N3)(O2)C2=CC=C(Cl)C=C2Cl)C=C1 |
| C35H38Cl2N8O4 | |
| MFCD00870168,MFCD00941396 | |
| Oriconazole; R51211 | |
| 1-(butan-2-yl)-4-{4-[4-(4-{[(2R,4S)-2-(2,4-dichlorophenyl)-2-[(1H-1,2,4-triazol-1-yl)methyl]-1,3-dioxolan-4-yl]methoxy}phenyl)piperazin-1-yl]phenyl}-4,5-dihydro-1H-1,2,4-triazol-5-one |
Specifications
| 84625-61-6 | |
| >110°C (230°F) | |
| MFCD00870168,MFCD00941396 | |
| Oriconazole; R51211 | |
| VHVPQPYKVGDNFY-ZPGVKDDISA-N | |
| 1-(butan-2-yl)-4-{4-[4-(4-{[(2R,4S)-2-(2,4-dichlorophenyl)-2-[(1H-1,2,4-triazol-1-yl)methyl]-1,3-dioxolan-4-yl]methoxy}phenyl)piperazin-1-yl]phenyl}-4,5-dihydro-1H-1,2,4-triazol-5-one | |
| 45039617 | |
| 98% |
| White | |
| C35H38Cl2N8O4 | |
| 14,5245 | |
| Insoluble in water; Solube in chloroform at 50mg/ml; Slightly soluble in ethanol or methanol | |
| CCC(C)N1N=CN(C1=O)C1=CC=C(C=C1)N1CCN(CC1)C1=CC=C(OC[C@H]2CO[C@@](CN3C=NC=N3)(O2)C2=CC=C(Cl)C=C2Cl)C=C1 | |
| 705.64 | |
| 705.63 | |
| Itraconazole |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'rtecsNumber' : XZ5481000
missing translation for 'tsca' : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.