missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITPRIPL1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-17003
This item is not returnable.
View return policy
Description
ITPRIPL1 Polyclonal specifically detects ITPRIPL1 in Human samples. It is validated for Western Blot.
Specifications
| ITPRIPL1 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| inositol 14,5-triphosphate receptor interacting protein-like 1, inositol 14,5-triphosphate receptor-interacting protein-like 1, KIAA1754L, KIAA1754-like | |
| Rabbit | |
| 63 kDa | |
| 0.1 mL | |
| Primary | |
| Human. Expected to cross react based on sequence identity: Mouse (89%), Rat (88%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 1mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q6GPH6 | |
| ITPRIPL1 | |
| Recombinant protein encompassing a sequence within the N-terminus region of human ITPRIPL1. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 150771 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction