missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ITPRIP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576541
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
This gene encodes a membrane-associated protein that binds the inositol 1,4,5-trisphosphate receptor (ITPR). The encoded protein enhances the sensitivity of ITPR to intracellular calcium signaling. Alternative splicing results in multiple transcript variants.
Specifications
| ITPRIP | |
| Polyclonal | |
| Unconjugated | |
| ITPRIP | |
| 4833424O12Rik; bA127L20; bA127L20.2; DANGER; inositol 1,4,5-triphosphate receptor interacting protein; inositol 1,4,5-triphosphate receptor-interacting protein; inositol 1,4,5-trisphosphate receptor interacting protein; inositol 1,4,5-trisphosphate receptor-interacting protein; ipri; Itprip; KIAA1754; mKIAA1754; Protein DANGER | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 414801, 85450 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q3TNL8, Q8IWB1 | |
| ITPRIP | |
| Recombinant fusion protein of human ITPRIP. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction