Learn More
ITPR3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA552021
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: ITSTKNEKIF QESIGLAIHL LDGGNTEIQK SFHNLMMSDK KSERFFKVLH DRMKRAQQET KSTVAVNMND LGSQPHEDRE PVDPTTKGRV ASFSIPGSSS RYSLGPSLRR GHEVSERVQS SEMGTSVLIM QPILRFLQLL CENHNRD Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 89%.
This gene encodes a receptor for inositol 1,4,5-trisphosphate, a second messenger that mediates the release of intracellular calcium. The receptor contains a calcium channel at the C-terminus and the ligand-binding site at the N-terminus. Knockout studies in mice suggest that type 2 and type 3 inositol 1,4,5-trisphosphate receptors play a key role in exocrine secretion underlying energy metabolism and growth.Specifications
ITPR3 | |
Polyclonal | |
Unconjugated | |
ITPR3 | |
inositol 1,4,5-triphosphate receptor 3; inositol 1,4,5-triphosphate receptor, type 3; inositol 1,4,5-trisphosphate receptor type 3; inositol 1,4,5-trisphosphate receptor, type 3; insP3R3; IP3 receptor; IP3 receptor isoform 3; IP3R; IP3R 3; Ip3r3; IP3R-3; IP3R3X; ITPR3; Itpr-3; Type 3 inositol 1,4,5-trisphosphate receptor; type 3 InsP3 receptor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3710 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q14573 | |
ITPR3 | |
Recombinant protein corresponding to Human ITPR3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only