missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ITPKB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111704
This item is not returnable.
View return policy
Description
Immunogen sequence: GGSPTLGLLG GSPSAQPGTG NVEAGIPSGR MLEPLPCWDA AKDLKEPQCP PGDRVGVQPG NSRVWQGTME KAGLAWTRGT GVQSEGTWES QR Highest antigen sequence indentity to the following orthologs: Mouse - 61%, Rat - 59%.
Inositol 1,4,5-trisphosphate (Ins(1,4,5)P3) regulates the level of calcium within the cell by releasing calcium from intracellular stores. Ins(1,4,5)P3 is phosphorylated by inositol 1,4,5-trisphosphate 3-kinase (IP3K) to form inositol 1,3,4,5-tetrakisphosphate (Ins(1,4,5)P4), which is is thought to regulate the influx of calcium across the plasma membrane. IP3K exists as three isoforms, IP3KA, B, and C. IP3KA, the most highly characterized isoform, is expressed in rat brain and testis. IP3KB is expressed in various rat tissues such as lung, thymus, testis, brain, and heart. IP3K activity is stimulated in the presence of calmodulin via phosphorylation by cAMP-dependent protein kinase, protein kinase C, or calcium/calmodulin dependent protein kinase II and, subsequently, mediates the inositol phosphate signaling pathways.
Specifications
| ITPKB | |
| Polyclonal | |
| Unconjugated | |
| ITPKB | |
| 1110033J02Rik; E130307H12Rik; Inositol 1,4,5-trisphosphate 3-kinase B; inositol-trisphosphate 3-kinase B; insP 3-kinase B; IP3 3-kinase B; IP3-3KB; IP3K; IP3K B; IP3KB; IP3K-B; Itpkb; PIG37; proliferation-inducing protein 37 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3707 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P27987 | |
| ITPKB | |
| Recombinant Protein corresonding to Human ITPKB. Recombinant protein control fragment (Product #RP-105573). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction