missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITPKB Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38060-100ul
This item is not returnable.
View return policy
Description
ITPKB Polyclonal antibody specifically detects ITPKB in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| ITPKB | |
| Polyclonal | |
| Western Blot 1:200 - 1:2000, ELISA | |
| EC 2.7.1.127, inositol 14,5-trisphosphate 3-kinase BIP3K, inositol-trisphosphate 3-kinase B, InsP 3-kinase B, IP3 3-kinase B, IP3-3KB, IP3K B, IP3KB, IP3K-B, PIG37, proliferation-inducing protein 37 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human ITPKB (NP_002212.3).,, Sequence:, MAVYCYALNSLVIMNSANEMKSGGGPGPSGSETPPPPRRAVLSPGSVFSPGRGASFLFPPAESLSPEEPRSPGGWRSGRRRLNSSSGSGSGSSGSSVSSPSWAGRLRGDRQQVVAAGTLSPPGPEEAKRKLRILQRELQNVQVNQKVGMFEAHIQAQSSAIQAPRSPRLGRARSPSPCPFRSSSQPPGRVLVQGARSEERRTKSWGEQCPETSGTDSGRKGGPSLCSSQVKKGMPPLPGRAAPTGSEAQGPSAFVRMEKG | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 3707 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction