missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ITPKA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578659
This item is not returnable.
View return policy
Description
Positive Control: mouse brain, rat brain Predicted Reactivity: Human (100%), Bovine (97%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Regulates inositol phosphate metabolism by phosphorylation of second messenger inositol 1,4,5-trisphosphate to Ins(1,3,4,5)P4. The activity of the inositol 1,4,5-trisphosphate 3-kinase is responsible for regulating the levels of a large number of inositol polyphosphates that are important in cellular signaling. Both calcium/calmodulin and protein phosphorylation mechanisms control its activity. It is also a substrate for the cyclic AMP-dependent protein kinase, calcium/calmodulin- dependent protein kinase II, and protein kinase C in vitro.
Specifications
| ITPKA | |
| Polyclonal | |
| Unconjugated | |
| ITPKA | |
| EC 2.7.1.127; inositol 1,4,5-triphosphate 3-kinase; inositol 1,4,5-trisphosphate 3-kinase A; inositol 145-triphosphate 3-kinase; inositol-trisphosphate 3-kinase A; insP 3-kinase A; IP3 3 kinase A; IP3 3-kinase A; IP3-3KA; IP3K A; IP3KA; IP3-kinase A; ITPKA | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 228550, 3706, 81677 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P17105, P23677, Q8R071 | |
| ITPKA | |
| Recombinant protein encompassing a sequence within the center region of human IP3KA. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction