missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITPK1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10568-1-AP-20UL
This item is not returnable.
View return policy
Description
ITPK1 encodes an enzyme that belongs to the inositol 1,3,4-trisphosphate 5/6-kinase family. This enzyme regulates the synthesis of inositol tetraphosphate, and downstream products, inositol pentakisphosphate and inositol hexakisphosphate. Inositol metabolism plays a role in the development of the neural tube. Disruptions in this gene are thought to be associated with neural tube defects. A pseudogene of this gene has been identified on chromosome X.Specifications
| ITPK1 | |
| Polyclonal | |
| Unconjugated | |
| ITPK1 | |
| Ins(1,3,4)P(3) 5/6 kinase, ITPK1, ITRPK1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 217837, 3705 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.26 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q13572, Q8BYN3 | |
| ITPK1 | |
| ITPK1 Fusion Protein Ag0834 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction