Learn More
Invitrogen™ ITPK1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597001
Description
Immunogen sequence: CVVQNFINHN AVLYKVFVVG ESYTVVQRPS LKNFSAGTSD RESIFFNSHN VSKPESSSVL TELDKIEGVF ERPSDEVIRE LSRALRQALG VSLFGIDIII NNQTGQHAVI DINAFPGYEG VSEFFTDLLN HIATVLQGQS TAMAATGDVA LLRHSKLLAE PAGGLVGERT CSASPGCCGS MMGQDAPWKA EADAGGTAKL PHQRLGCNAG VSPSFQQHCV ASLATKASSQ; Positive Samples: HeLa, HepG2, Mouse testis, Mouse brain, Mouse intestine, Rat brain.
ITPK1 encodes an enzyme that belongs to the inositol 1,3,4-trisphosphate 5/6-kinase family. This enzyme regulates the synthesis of inositol tetraphosphate, and downstream products, inositol pentakisphosphate and inositol hexakisphosphate. Inositol metabolism plays a role in the development of the neural tube. Disruptions in this gene are thought to be associated with neural tube defects. A pseudogene of this gene has been identified on chromosome X.
Specifications
| ITPK1 | |
| Polyclonal | |
| Unconjugated | |
| ITPK1 | |
| BC031182; inositol 1,3,4-triphosphate 5/6 kinase; inositol 1,3,4-trisphosphate 5/6-kinase; Inositol-tetrakisphosphate 1-kinase; inositol-triphosphate 5/6-kinase; ins(1,3,4)P(3) 5/6-kinase; ITPK1; ITRPK1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 217837, 3705, 500709 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.68 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q13572, Q8BYN3 | |
| ITPK1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 185-414 of human ITPK1 (NP_0550312). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.