missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITPA Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£161.00 - £375.00
Specifications
| Antigen | ITPA |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18095676
|
Novus Biologicals
NBP3-04495-20UL |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18343057
|
Novus Biologicals
NBP3-04495-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ITPA Polyclonal specifically detects ITPA in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
| ITPA | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 3704 | |
| IgG | |
| Affinity purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| C20orf37, dJ794I6.3, EC 3.6.1, EC 3.6.1.19, HLC14-06-P, Inosine triphosphatase, inosine triphosphatase (nucleoside triphosphate pyrophosphatase), inosine triphosphatase-A, inosine triphosphate pyrophosphatase, inosine triphosphate pyrophosphohydrolase, ITPase, My049 protein, nucleoside triphosphate diphosphatase, Putative oncogene protein hlc14-06-p | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-194 of human ITPA (NP_258412.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title