Learn More
ITPA Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA554614
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: GFEDKSAYAL CTFALSTGDP SQPVRLFRGR TSGRIVAPRG CQDFGWDPCF QPDGYEQTYA EMPKAEKNAV SHRFRALLEL QEYFGSLAA Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 83%.
Pyrophosphatase that hydrolyzes the non-canonical purine nucleotides inosine triphosphate (ITP), deoxyinosine triphosphate (dITP) as well as 2'-deoxy-N-6-hydroxylaminopurine triposphate (dHAPTP) and xanthosine 5'-triphosphate (XTP) to their respective monophosphate derivatives. The enzyme does not distinguish between the deoxy- and ribose forms. Probably excludes non-canonical purines from RNA and DNA precursor pools, thus preventing their incorporation into RNA and DNA and avoiding chromosomal lesions.Specifications
ITPA | |
Polyclonal | |
Unconjugated | |
ITPA | |
2010016I08Rik; AU020102; C20orf37; dJ794I6.3; HAMAP-Rule:MF_03148}; HLC14-06-P; inosine triphosphatase; inosine triphosphatase (nucleoside triphosphate pyrophosphatase); inosine triphosphatase {ECO:0000255; inosine triphosphate pyrophosphatase; inosine triphosphate pyrophosphatase {ECO:0000255; inosine triphosphate pyrophosphohydrolase; Itp; ITPA; ITPase; ITPase {ECO:0000255; My049; My049 protein; Non-canonical purine NTP pyrophosphatase; non-canonical purine NTP pyrophosphatase {ECO:0000255; non-standard purine NTP pyrophosphatase; non-standard purine NTP pyrophosphatase {ECO:0000255; NTPase; NTPase {ECO:0000255; nucleoside triphosphate pyrophosphatase; nucleoside-triphosphate diphosphatase; nucleoside-triphosphate diphosphatase {ECO:0000255; Nucleoside-triphosphate pyrophosphatase; nucleoside-triphosphate pyrophosphatase {ECO:0000255; OK/SW-cl.9; Putative oncogene protein hlc14-06-p | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3704 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BY32 | |
ITPA | |
Recombinant protein corresponding to Human ITPA | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only