missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ITM2C Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5148930
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Negative regulator of beta amyloid peptide production. May inhibit the processing of APP by blocking its access to alpha-and beta-secretase. Binding to the beta-secretase-cleaved APP C-terminal fragment is negligible, suggesting that ITM2C is a poor gamma-secretase cleavage inhibitor. May play a role in TNF-induced cell death and neuronal differentiation.
Specifications
| ITM2C | |
| Polyclonal | |
| Unconjugated | |
| ITM2C | |
| 3110038L02Rik; BRI3; Bricd2c; BRICHOS domain containing 2C; cerebral protein 14; CT-BRI3; E25; E25C; hRPC.1050_D_4; hucep-14; Integral membrane protein 2C; integral membrane protein 3; Itm2c; ITM3; NPD018; PSEC0047; Transmembrane protein BRI3 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 81618 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9NQX7 | |
| ITM2C | |
| E. coli-derived recombinant human ITM2C, Ala86-Val267. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction