missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITM2C Mouse anti-Human, Clone: 2E8G11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60095-1-IG-20UL
This item is not returnable.
View return policy
Description
Negative regulator of beta amyloid peptide production. May inhibit the processing of APP by blocking its access to alpha-and beta-secretase. Binding to the beta-secretase-cleaved APP C-terminal fragment is negligible, suggesting that ITM2C is a poor gamma-secretase cleavage inhibitor. May play a role in TNF-induced cell death and neuronal differentiation.Specifications
| ITM2C | |
| Monoclonal | |
| 0.95 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9NQX7 | |
| ITM2C | |
| ITM2C Fusion Protein Ag4409 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunocytochemistry, Western Blot, Flow Cytometry, Immunofluorescence | |
| 2E8G11 | |
| Unconjugated | |
| ITM2C | |
| BRICD2C, Cerebral protein 14, E25, E25C, integral membrane protein 2C, ITM2C, ITM3, PSEC0047, Transmembrane protein BRI3 | |
| Mouse | |
| Caprylic Acid Precipitation | |
| RUO | |
| 81618 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction