missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ITM2B Polyclonal Antibody

Product Code. 16380035
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16380035 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16380035 Supplier Invitrogen™ Supplier No. PA583079

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: YFALQPDDVY YCGIKYIKDD VILNEPSADA PAALYQTIEE NIKIFEEEEV EFISVPVPEF ADSDPANIVH DFNKKLTAYL DLNLDKCYVI PLNTSIVMPP RNLLELLINI KAGTYLPQSY LIHEHMVITD RIENIDHLGF FIYRLCH Highest antigen sequence indentity to the following orthologs: Mouse - 95%, Rat - 96%.

Amyloid precursor proteins are processed by beta-secretase and gamma-secretase to produce beta-amyloid peptides which form the characteristic plaques of Alzheimer disease. This gene encodes a transmembrane protein which is processed at the C-terminus by furin or furin-like proteases to produce a small secreted peptide which inhibits the deposition of beta-amyloid. Mutations which result in extension of the C-terminal end of the encoded protein, thereby increasing the size of the secreted peptide, are associated with two neurogenerative diseases, familial British dementia and familial Danish dementia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ITM2B
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ITM2B
Gene Accession No. Q9Y287
Gene Alias ABRI; ABri/ADan amyloid peptide; ABri23; AI256040; Bri; BRI2; BRI2 ICD; BRI2 intracellular domain; BRI2, membrane form; Bri2-23; Bri23 peptide; BRI2C, soluble form; Bricd2b; BRICHOS domain containing 2B; C-terminal peptide; D14Sel6; E25B; E25BMM; E3-16; FBD; imBRI2; Immature BRI2; integral membrane protein 2B; Integral membrane protein 2B (E25B protein); ITM2B; Mature BRI2; mBRI2; OTTHUMP00000018394; P23 peptide; Protein E25B; RDGCA; RP11-196P14.1; Transmembrane protein BRI
Gene Symbols ITM2B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ITM2B. Recombinant protein control fragment (Product #RP-102188).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9445
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.