missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ITM2B Monoclonal Antibody (OTI1C11)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525513
This item is not returnable.
View return policy
Description
ITM2B Monoclonal Antibody for Western Blot, IHC (P), Flow
Amyloid precursor proteins are processed by beta-secretase and gamma-secretase to produce beta-amyloid peptides which form the characteristic plaques of Alzheimer disease. This gene encodes a transmembrane protein which is processed at the C-terminus by furin or furin-like proteases to produce a small secreted peptide which inhibits the deposition of beta-amyloid. Mutations which result in extension of the C-terminal end of the encoded protein, thereby increasing the size of the secreted peptide, are associated with two neurogenerative diseases, familial British dementia and familial Danish dementia.
Specifications
| ITM2B | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9Y287 | |
| ITM2B | |
| Full length human recombinant protein of ITM2B produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI1C11 | |
| Unconjugated | |
| ITM2B | |
| ABRI; ABri/ADan amyloid peptide; ABri23; AI256040; Bri; BRI2; BRI2 ICD; BRI2 intracellular domain; BRI2, membrane form; Bri2-23; Bri23 peptide; BRI2C, soluble form; Bricd2b; BRICHOS domain containing 2B; C-terminal peptide; D14Sel6; E25B; E25BMM; E3-16; FBD; imBRI2; Immature BRI2; integral membrane protein 2B; Integral membrane protein 2B (E25B protein); ITM2B; Mature BRI2; mBRI2; OTTHUMP00000018394; P23 peptide; Protein E25B; RDGCA; RP11-196P14.1; Transmembrane protein BRI | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9445 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction