missing translation for 'onlineSavingsMsg'
Learn More

ITM2B Antibody [DyLight 650], Novus Biologicals™

Product Code. 30736748 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30736748 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30736748 Supplier Novus Biologicals Supplier No. AF7015W

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

ITM2B Polyclonal antibody specifically detects ITM2B in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen ITM2B
Applications Immunohistochemistry
Classification Polyclonal
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias ABRI, ABri/ADan amyloid peptide, BRI2, BRICD2B, BRICHOS domain containing 2B, BRIFBD, E25B, E3-16, integral membrane protein 2B, Protein E25B, Transmembrane protein BRI
Host Species Sheep
Immunogen E. coli-derived recombinant human ITM2B, Ala60-Ser266, Accession # Q9Y287
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9445
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.