missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ITM2A Polyclonal specifically detects ITM2A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | ITM2A |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | DyLight 350 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | BRICHOS domain containing 2A, E25ABRICD2A, integral membrane protein 2A, Protein E25 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human ITM2A. |
| Purification Method | Antigen Affinity-purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?