Learn More
ITL BioMedical Safety SubCulture Unit
Product Code. 15235636
Encompass_Preferred
Change view
Click to view available options
Product Type:
Safety Subculture Unit
Unit Size:
Pack of 60
Product Code. Product Type unitSize
15235636 Safety Subculture Unit Pack of 60
1 options
Product Code. 15235636 Supplier ITL BioMedical Supplier No. A100720
Only null left
Add to basket
Add to basket
Needleless safety device for subculturing blood culture bottles.
Used to transfer sample from positive blood culture bottles for further testing, bringing a new level of safety to microbiology labs.
  • Easy to use and versatile
  • Two procedure options to obtain samples for further testing: drop sample method and syringe method
  • Neither method requires needle which reduces the risk of potential needle stick injury
  • Does not contain latex
  • Compatible fit with various culture bottles
  • Limits exposure to gas or culture discharge
  • Single use only
Specifications
Product Type Safety Subculture Unit