missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ITK, GST Tag Recombinant Protein
ITK Recombinant Human Protein
£434.00 - £6900.00
Specifications
Accession Number | NP_005537 |
---|---|
Gene Alias | ITK, EMT, LYK, PSCTK2 |
Protein Tag | GST Tag |
Expression System | Baculovirus |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11523607
|
Invitrogen
PV3875 |
10μg |
£434.00
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10654613
|
Invitrogen
PV4193 |
1mg |
£6900.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10348532
|
Invitrogen
PR6992A |
£1602.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
ITK is a tyrosine kinase expressed in T-cells which contains both SH2 and SH3 domains. ITK is thought to play a role in T-cell proliferation and differentiation.Specifications
NP_005537 | |
ITK, EMT, LYK, PSCTK2 | |
GST Tag | |
Baculovirus |
For Research Use Only. Not for use in diagnostic procedures.