missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eurofins DiscoverX Itk Protein, Active
Click to view available options
Quantity:
10 μg
250 μg
Unit Size:
10µg
250µg
Description
ITK (Interleukin-2-inducible T-cell kinase) is an important member of the Tec family of non-receptor tyrosine kinases that play a central role in signaling through antigen receptors such as the T-cell receptor, B-cell receptor, and Fce.
GenBank NM_005546
Specifications
Specifications
| Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Molecular Weight (g/mol) | 34 kDa |
| Quantity | 250 μg |
| Species | Human |
| Recombinant | Recombinant |
| Protein Tag | N-terminal 6His |
| Expression System | Recombinant enzyme expressed in Sf21 insect cells |
| Protein Form | Truncated |
| Purity or Quality Grade | ≥72% |
| Protein | Itk, active |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction