missing translation for 'onlineSavingsMsg'
Learn More

ITK Antibody (8F10), Novus Biologicals™

Product Code. 18358574 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18358574 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18358574 Supplier Novus Biologicals Supplier No. NBP315208

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

ITK Monoclonal antibody specifically detects ITK in Human samples. It is validated for ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen ITK
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone 8F10
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry 1:50-1:200
Formulation PBS (pH 7.4), 150mM NaCl, and 50% glycerol.
Gene Alias EC 2.7.10, EC 2.7.10.2, EMTMGC126257, homolog of mouse T-cell itk/tsk, IL2-inducible T-cell kinase, Kinase EMT, LYKMGC126258, PSCTK2, T-cell-specific kinase, tyrosine-protein kinase ITK/TSK, Tyrosine-protein kinase Lyk
Host Species Rabbit
Immunogen A synthesized peptide derived from human ITK. UniProt ID (Q08881).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 3702
Target Species Human
Content And Storage Store at -20 to -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.