missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ITIH4 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142437
This item is not returnable.
View return policy
Description
Additional Information: The immunizing peptide represents a region of the 35kDa chain. This chain is glycosylated giving rise to an apparent higher molecular weight. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.
The inter-alpha-trypsin inhibitor (ITI, I-alpha-I) family is a classic example of protein-glycosaminoglycoprotein (PGP) complexes. They occur constitutively in plasma at relatively high concentrations and results from alternating combinations of three kinds of heavy chains with a common light chain called the bikunin proteoglycan. Bikunin has two proteinase inhibotor domains and belongs to the Kunitz-type protease inhibitor family; it displays an inhibitory activity against trypsin, leukocyte elastase and plasmin. The heavy chains do not have any protease inhibitory properties but have the capacity to interact in vitro and in vivo with hyaluronic acid and this binding promotes the stability of the extra-cellular matrix. The ITI protein family is suspected of playing a key role in the extra-cellular matrix biology.
Specifications
| ITIH4 | |
| Polyclonal | |
| Unconjugated | |
| ITIH4 | |
| 35 kDa inter-alpha-trypsin inhibitor heavy chain H4; 70 kDa inter-alpha-trypsin inhibitor heavy chain H4; DKFZp686G21125; Gp120; H4P; IHRP; Inter alpha-trypsin inhibitor, heavy chain 4; inter-alpha (globulin) inhibitor H4 (plasma Kallikrein-sensitive glycoprotein); inter-alpha-inhibitor heavy chain 4; inter-alpha-trypsin inhibitor family heavy chain-related protein; inter-alpha-trypsin inhibitor heavy chain 4; inter-alpha-trypsin inhibitor heavy chain family member 4; inter-alpha-trypsin inhibitor heavy chain family, member 4; inter-alpha-trypsin inhibitor heavy chain H4; inter-alpha-trypsin inhibitor, heavy chain-like, 1; ITI heavy chain H4; Itih4; Itih-4; ITI-HC4; ITIHL1; PK120; PK-120; Plasma kallikrein sensitive glycoprotein 120; plasma kallikrein-sensitive glycoprotein 120; PRO1851 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 3700 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q14624 | |
| ITIH4 | |
| Peptide with sequence ERLCVDPRHRQGPVN. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction