missing translation for 'onlineSavingsMsg'
Learn More

ITIH4 Antibody, Novus Biologicals™

Product Code. 18417100 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25 μL
Unit Size:
0.10mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18417100 25 μL 25µL
18289055 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18417100 Supplier Novus Biologicals Supplier No. NBP18177725ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ITIH4 Polyclonal specifically detects ITIH4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ITIH4
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.4 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:500-1:1000
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q14624
Gene Alias DKFZp686G21125, H4P, heavy chain-like, 1, IHRP, inter-alpha (globulin) inhibitor H4 (plasma Kallikrein-sensitive glycoprotein), inter-alpha-trypsin inhibitor family heavy chain-related protein, inter-alpha-trypsin inhibitor heavy chain H4, ITI-HC4, ITIHL1, PK-120, plasma kallikrein-sensitive glycoprotein 120, PRO1851
Gene Symbols ITIH4
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:LAYSFVTPLTSMVVTKPDDQEQSQVAEKPMEGESRNRNVHSAGAAGSRMNFRPGVLSSRQLGLPGPPDVPDHAAYHPFRRLAILPASAPPATSNPDPAVSRVMNMKIEETTMTTQTPA
Molecular Weight of Antigen 103 kDa
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3700
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.