missing translation for 'onlineSavingsMsg'
Learn More

ITIH4 Antibody [mFluor Violet 450 SE], Novus Biologicals™

Product Code. 30735289 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30735289 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735289 Supplier Novus Biologicals Supplier No. AF8157MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

ITIH4 Polyclonal antibody specifically detects ITIH4 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ITIH4
Applications Western Blot
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Formulation 50mM Sodium Borate
Gene Alias DKFZp686G21125, H4P, heavy chain-like, 1, IHRP, inter-alpha (globulin) inhibitor H4 (plasma Kallikrein-sensitive glycoprotein), inter-alpha-trypsin inhibitor family heavy chain-related protein, inter-alpha-trypsin inhibitor heavy chain H4, ITI-HC4, ITIHL1, PK-120, plasma kallikrein-sensitive glycoprotein 120, PRO1851
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human ITIH4, Glu29-Leu930 (Ile85Asn, Pro698Thr), Accession # Q14624
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3700
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.