missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ITIH3 Monoclonal specifically detects ITIH3 in Human samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | ITIH3 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 49 |
| Conjugate | Alexa Fluor 405 |
| Dilution | ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | H3 polypeptide, inter-alpha (globulin) inhibitor H3, inter-alpha (globulin) inhibitor, H3 polypeptide, inter-alpha-inhibitor heavy chain 3, inter-alpha-trypsin inhibitor heavy chain H3, ITI heavy chain H3, ITI-HC3, serum-derived hyaluronan-associated protein, SHAP |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human ITIH3 (NP_002208.3; Met1-Asp651). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?