missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ITIH1 (Cleaved Asp672) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA537430
This item is not returnable.
View return policy
Description
This antibody detects endogenous protein at a molecular weight of 71 and 101 kDa. Purity is >95% by SDS-PAGE.
The inter-alpha trypsin inhibitor (ITI) family is a group of structurally related plasma serine protease inhibitors synthesized in the liver and built up from different combinations of three highly homologous heavy chains (ITI-H1, ITI-H2 and ITI-H3) and one light chain (Bikunin). A fourth member of the ITI family, ITI-H4 (also known as I a IH4P) harbors a Pro-rich region (PRR) in its C-terminus. ITI is a glycoprotein composed of three polypeptides linked by chondroitin sulphate: two heavy chains, ITI-H1 and ITI-H2, and Bikunin. Bikunin confers the protease-inhibitor function of ITI. The heavy chains of the ITI family, designated as SHAPs (for serum-derived hyaluronan-associated proteins), bind covalently to hyaluronic acid (HA), resulting in pericellular matrix stabilization. ITI-H1 contains a potential peptide which could stimulate a broad spectrum of phagocytotic cells. Although ITI-H1, ITI-H3 and Bikunin have antitumoral and antimetastatic properties in the cell, they are also associated with malignant transformation of lung tissue. ITI-H1 and ITI-H2 are associated with calcium oxalate stone formation in kidney and urine.
Specifications
| ITIH1 (Cleaved Asp672) | |
| Polyclonal | |
| Unconjugated | |
| ITIH1 | |
| H1P; IATIH; IGHEP1; inter-alpha (globulin) inhibitor, H1 polypeptide; inter-alpha trypsin inhibitor, heavy chain 1; inter-alpha-inhibitor heavy chain 1; inter-alpha-trypsin inhibitor complex component III; inter-alpha-trypsin inhibitor heavy chain 1; inter-alpha-trypsin inhibitor heavy chain H1; Intin1; ITI heavy chain H1; ITIH; ITIH1; Itih-1; ITI-HC1; Serum-derived hyaluronan-associated protein; SHAP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 16424, 306251, 3697 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P19827, Q61702 | |
| ITIH1 | |
| Synthetic peptide corresponding to amino acids 650-700 of human ITI-H1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction