missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ITIH1 Polyclonal antibody specifically detects ITIH1 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | ITIH1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot |
| Formulation | PBS (pH 7.4) |
| Gene Accession No. | NP_002206.1 |
| Gene Alias | H1P, IATIH, IGHEP1, inter-alpha (globulin) inhibitor H1, inter-alpha (globulin) inhibitor, H1 polypeptide, Inter-alpha-inhibitor heavy chain 1, Inter-alpha-trypsin inhibitor complex component III, inter-alpha-trypsin inhibitor heavy chain H1, ITI heavy chain H1, ITIH, ITI-HC1, MGC126415, Serum-derived hyaluronan-associated protein, SHAP |
| Host Species | Rabbit |
| Immunogen | ITIH1 (NP_002206.1, 1 a.a. - 911 a.a.) full-length human protein. MDGAMGPRGLLLCMYLVSLLILQAMPALGSATGRSKSSEKRQAVDTAVDGVFIRSLKVNCKVTSRFAHYVVTSQVVNTANEAREVAFDLEIPKTAFISDFAVTADGNAFIGDIKDKVTAWKQYRKAAISGENAGLVRASGRTMEQFTIHLTVNPQSKVTFQLTYEEVLKRNHMQYEIVIKVKPKQLVHHFEIDVDIFEPQGISKLDAQASFLPKELAAQTIKKSFSGKKGHVLFRPTVSQQQSCPTCSTSLLNGHFKVTYDVSRDKICDLLVANNHFAHFFAPQNLTNMNKNVVFVIDISGSMRGQKVKQTKEALLKILGDMQPGDYFDLVLFGTRVQSWKGSLVQASEANLQAAQDFVRGFSLDEATNLNGGLLRGIEILNQVQESLPELSNHASILIMLTDGDPTEGVTDRSQILKNVRNAIRGRFPLYNLGFGHNVDFNFLEVMSMENNGRAQRIYEDHDATQQLQGFYSQVAKPLLVDVDLQYPQDAVLALTQNHHKQYYEGSEIVVAGRIADNKQSSFKADVQAHGEGQEFSITCLVDEEEMKKLLRERGHMLENHVERLWAYLTIQELLAKRMKVDREVRANLSSQALRMSLDYGFVTPLTSMSIRGMADQDGLKPTIDKPSEDSPPLEMLGPRRTFVLSALQPSPTHSSSNTQRLPDRVTGVDTDPHFIIHVPQKEDTLCFNINEEPGVILSLVQDPNTGFSVNGQLIGNKARSPGQHDGTYFGRLGIANPATDFQLEVTPQNITLNPGFGGPVFSWRDQAVLRQDGVVVTINKKRNLVVSVDDGGTFEVVLHRVWKGSSVHQDFLGFYVLDSHRMSARTHGLLGQFFHPIGFEVSDIHPGSDPTKPDATMVVRNRRLTVTRGLQKDYSKDPWHGAEVSCWFIHNNGAGLIDGAYTDYIVPDIF |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?