missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITIH1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-81783
This item is not returnable.
View return policy
Description
ITIH1 Polyclonal specifically detects ITIH1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| ITIH1 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| H1P, IATIH, IGHEP1, inter-alpha (globulin) inhibitor H1, inter-alpha (globulin) inhibitor, H1 polypeptide, Inter-alpha-inhibitor heavy chain 1, Inter-alpha-trypsin inhibitor complex component III, inter-alpha-trypsin inhibitor heavy chain H1, ITI heavy chain H1, ITIH, ITI-HC1, MGC126415, Serum-derived hyaluronan-associated protein, SHAP | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 3697 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| ITIH1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:QPGDYFDLVLFGTRVQSWKGSLVQASEANLQAAQDFVRGFSLDEATNLNGGLLRGIEILNQVQESLPELSNHA | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction