missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITI-214, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Description
ITI-214 is a potent, CNS-active, orally bioavailable PDE1 inhibitor (Ki of 58 pM) with excellent selectivity against other PDE family members and against a panel of enzymes, receptors, transporters and ion channels. ITI-214 inhibits recombinant full-length human PDE1A, PDE1B and PDE1C with Kis of 33 pM, 380 pM and 35 pM, respectively. ITI-214 shows efficacy in various animal models of motor and cognitive functions.
Specifications
Specifications
| Chemical Name or Material | ITI-214 |
| CAS | 1642303-38-5 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C29H29FN7O5P |
| Quantity | 50 mg |
| Solubility Information | DMSO : ≥ 30 mg/mL (49.54 mM) |
| SMILES | O=C(C1=C(NC2=CC=CC=C2)N(CC3=CC=C(C4=NC(F)=CC=C4)C=C3)N=C1N56)N(C)C5=N[C@@]7([H])[C@]6([H])CCC7.O=P(O)(O)O |
| Molecular Weight (g/mol) | 605.56 |
| Formula Weight | 605.56 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction