missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITI-214 free base, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Description
ITI-214 free base inhibits recombinant full-length human PDE1A, PDE1B and PDE1C with Kis of 33 pM, 380 pM and 35 pM, respectively. ITI-214 free base shows efficacy in various animal models of motor and cognitive functions.
Specifications
Specifications
| Chemical Name or Material | ITI-214 free base |
| CAS | 1160521-50-5 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C29H26FN7O |
| Quantity | 5 mg |
| SMILES | O=C(C1=C(NC2=CC=CC=C2)N(CC3=CC=C(C4=NC(F)=CC=C4)C=C3)N=C1N56)N(C)C5=N[C@@]7([H])[C@]6([H])CCC7 |
| Molecular Weight (g/mol) | 507.56 |
| Formula Weight | 507.56 |
| Percent Purity | 98.0% |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction