missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ITGBL1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA552179

Product Code. 15750131

  • £471.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: VCGECTCHDV DPTGDWGDIH GDTCECDERD CRAVYDRYSD DFCSGHGQCN CGRCDCKAGW YGKKCEHPQS CTLSAEESIR KCQGSSDLPC SGRGKCECGK CTCYPPGDRR VYGKTCECDD RRCEDLDGV Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 93%.

The protein encoded by this gene is a component of the F-type ATPase found in the mitochondrial matrix. F-type ATPases are composed of a catalytic core and a membrane proton channel. The encoded protein appears to be part of the connector linking these two components and may be involved in transmission of conformational changes or proton conductance.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ITGBL1
Polyclonal
Unconjugated
ITGBL1
B930011D01Rik; integrin beta like 1; integrin beta-like protein 1; integrin subunit beta like 1; integrin, beta-like 1; integrin, beta-like 1 (with EGF-like repeat domains); ITGBL1; OSCP; Osteoblast-specific cysteine-rich protein; ten integrin EGF-like repeat domain-containing protein; TIED; with EGF-like repeat domains
Rabbit
Antigen affinity chromatography
RUO
9358
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O95965
ITGBL1
Recombinant protein corresponding to Human ITGBL1. Recombinant protein control fragment (Product #RP-88644).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.