missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITGB8 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA555691
This item is not returnable.
View return policy
Description
Immunogen sequence: RIHNQCSDYN LDCMPPHGYI HVLSLTENIT EFEKAVHRQK ISGNIDTPEG GFDAMLQAAV CESHIG Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
ITGB8 (Integrin beta 8) is a member of the integrin beta chain family and is a single-pass type I membrane protein with a VWFA domain and four cysteine-rich repeats. This protein noncovalently binds to an alpha subunit to form a heterodimeric integrin complex. In general, integrin complexes mediate cell-cell and cell-extracellular matrix interactions and this complex plays a role in human airway epithelial proliferation. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized.
Specifications
| ITGB8 | |
| Polyclonal | |
| Unconjugated | |
| ITGB8 | |
| ITGB8, integrin beta-8, integrin, beta 8 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3696 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P26012 | |
| ITGB8 | |
| Recombit protein corresponding to Human ITGB8 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction