missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ITGB5 Recombinant Rabbit Monoclonal Antibody (K01_1S98)

Product Code. 30828983
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30828983 50 μL
30826959 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30828983 Supplier Invitrogen™ Supplier No. MA564931
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

ITGB5 Recombinant Monoclonal Antibody for Western Blot

Integrins are important extracellular matrix (ECM) receptor proteins located on cell surfaces. They are hetrodimers composed of an alpha and a beta transmembrane glycoprotein subunit. Around 22 different integrins (different alpha/ beta subunit combinations) are found in nature. Integrins are generally present in high concentrations at the cell surface, but, unlike most other cell-surface receptors, they bind ligands with very low affinity. Due to their weak individual binding, integrins need to cluster and bind in-groups in order to effectively bind the ECM. Integrins bind many different ligands including laminin. Each integrin is made up of a large N-terminal extracellular domain that binds the ECM ligand and a small C-terminal cytoplasmic domain that mediates interaction with the actin cytoskeleton and signaling function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ITGB5
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1S98
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 0.05% BSA, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene Itgb5
Gene Accession No. P18084
Gene Alias [b]5; [b]-5; [b]5A; [b]5B; AA475909; AI874634; beta5; beta-5; CD61; ESTM23; FLJ26658; GP3A; GPIIIa; I79_000655; integrin beta 5; integrin beta-5; integrin subunit beta 5; integrin, beta 5; ITGB5; RGD1563276; testis secretory sperm-binding protein Li 217p
Gene Symbols Itgb5
Host Species Rabbit
Immunogen Recombinant protein of human Integrin beta 5.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100765378, 257645, 3693
Target Species Human, Rat, Hamster
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.