missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ITGB1BP3 Antibody (OS2966) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
£162.00 - £695.00
Specifications
| Antigen | ITGB1BP3 |
|---|---|
| Clone | OS2966 |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30046524
|
Novus Biologicals™
NBP3-28236-1mg |
1 mg |
£695.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30047882
|
Novus Biologicals™
NBP3-28236-100ug |
100 μg |
£266.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30047515
|
Novus Biologicals™
NBP3-28236-50ug |
50 μg |
£162.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ITGB1BP3 Monoclonal antibody specifically detects ITGB1BP3 in Human samples. It is validated for Flow Cytometry,ELISA,Functional AssaySpecifications
| ITGB1BP3 | |
| Flow Cytometry, ELISA, Functional Assay | |
| Unconjugated | |
| Human | |
| Cancer, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Mesenchymal Stem Cell Markers, Neuronal Cell Markers, Signal Transduction, Stem Cell Markers | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| EC 2.7.1.22, EC 2.7.1.n4, integrin beta 1 binding protein 3, Integrin beta-1-binding protein 3, MGC126624, MIBPNRK 2, Muscle integrin-binding protein, muscle-specific beta 1 integrin binding protein, nicotinamide riboside kinase 2, Nicotinic acid riboside kinase 2, NmR-K 2, NRK2RNK 2, Ribosylnicotinamide kinase 2, Ribosylnicotinic acid kinase 2 | |
| Integrin b1/ ITGB1/ CD29 | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. |
| OS2966 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P05556 | |
| 27231 | |
| IgG1 | |
| Protein A purified | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title