missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ITGB1BP2 Monoclonal Antibody (3G9)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA521660
This item is not returnable.
View return policy
Description
Peptide Sequence: KASQTELHVH IVFDGNRVFQ AQMKLWGVIN VEQSSVFLMP SRVEISLVKA DPGSWAQLEH PDALAKKARA GVVL.
Integrin beta-1 binding protein 2 (ITGB1BP2), commonly referred to as Melusin, is a gene located on human chromosome 8p21.3. ITGB1BP2 is known for its role in cardiac muscle function, encoding a protein that interacts with integrin β1, which is essential for integrin signaling pathways. This gene is implicated in hypertrophic cardiomyopathy and dilated cardiomyopathy, conditions that affect heart muscle structure and function. ITGB1BP2 gene variations have been observed in families with hypertension or primary hypertrophic cardiomyopathy, although clear genotype/phenotype correlations were not conclusively established. Notably, a specific missense mutation in ITGB1BP2, involving alanine-to-glycine substitution at position 313, has been identified in patients with dilated cardiomyopathy. Melusin, the protein encoded by ITGB1BP2, plays a crucial role in mechanotransduction processes, influencing cellular responses to mechanical stress, which is particularly important in cardiac tissue adaptation and repair mechanisms.
Specifications
| ITGB1BP2 | |
| Monoclonal | |
| 0.32 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q9UKP3 | |
| ITGB1BP2 | |
| ITGB1BP2 (NP_036410, 246 a.a. ~ 319 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 3G9 | |
| Unconjugated | |
| ITGB1BP2 | |
| Chordc3; integrin beta 1 binding protein (melusin) 2; integrin beta 1 binding protein 2; integrin beta-1-binding protein 2; integrin subunit beta 1 binding protein 2; ITGB1BP; Itgb1bp2; Melusin; MGC119214; MSTP015; RGD1565015 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 26548 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction