missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ITGB1BP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584755
This item is not returnable.
View return policy
Description
Immunogen sequence: VSKYGIKVST SDQYDVLHRH ALYLIIRMVC YDDGLGAGKS LLALKTTDAS NEEYSLWVYQ CNSLEQAQAI CKVLSTAFDS VLT Highest antigen sequence indentity to the following orthologs: Rat - 96%, Mouse - 96%.
Key regulator of the integrin-mediated cell-matrix interaction signaling by binding to the ITGB1 cytoplasmic tail and preventing the activation of integrin alpha-5/beta-1 (heterodimer of ITGA5 and ITGB1) by talin or FERMT1. Plays a role in cell proliferation, differentiation, spreading, adhesion and migration in the context of mineralization and bone development and angiogenesis. Stimulates cellular proliferation in a fibronectin-dependent manner. Involved in the regulation of beta-1 integrin-containing focal adhesion (FA) site dynamics by controlling its assembly rate during cell adhesion; inhibits beta-1 integrin clustering within FA by directly competing with talin TLN1, and hence stimulates osteoblast spreading and migration in a fibronectin-and/or collagen-dependent manner. Acts as a guanine nucleotide dissociation inhibitor (GDI) by regulating Rho family GTPases during integrin-mediated cell matrix adhesion; reduces the level of active GTP-bound form of both CDC42 and RAC1 GTPases upon cell adhesion to fibronectin. Stimulates the release of active CDC42 from the membranes to maintain it in an inactive cytoplasmic pool. Participates in the translocation of the Rho-associated protein kinase ROCK1 to membrane ruffles at cell leading edges of the cell membrane, leading to an increase of myoblast cell migration on laminin. Plays a role in bone mineralization at a late stage of osteoblast differentiation; modulates the dynamic formation of focal adhesions into fibrillar adhesions, which are adhesive structures responsible for fibronectin deposition and fibrillogenesis. Plays a role in blood vessel development; acts as a negative regulator of angiogenesis by attenuating endothelial cell proliferation and migration, lumen formation and sprouting angiogenesis by promoting AKT phosphorylation and inhibiting ERK1/2 phosphorylation through activation of the Notch signaling pathway. Promotes transcriptional activity of the MYC promoter.
Specifications
| ITGB1BP1 | |
| Polyclonal | |
| Unconjugated | |
| ITGB1BP1 | |
| AI449260; AU019480; bodenin; DKFZp686K08158; Icap1; ICAP-1; ICAP1A; ICAP-1A; ICAP-1alpha; ICAP1B; ICAP-1B; integrin beta 1 binding protein 1; integrin beta-1-binding protein 1; Integrin cytoplasmic domain-associated protein 1; integrin cytoplasmic domain-associated protein 1-alpha; integrin cytoplasmic domain-associated protein 1-beta; integrin subunit beta 1 binding protein 1; ITGB1BP1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9270 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O14713 | |
| ITGB1BP1 | |
| Recombinant protein corresponding to Human ITGB1BP1. Recombinant protein control fragment (Product #RP-108528). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction