missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITGB1 Monoclonal Antibody (MEM-101A), FITC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen CD2901
Code : 50
Additional Details : Weight : 0.50000kg
Description
Commonly used, FITC conjugates provide relatively high absorptivity, excellent fluorescence quantum yield, and good water solubility.
ITGB1 Monoclonal antibody specifically detects ITGB1 in Human samples. It is validated for Flow Cytometry.Specifications
ITGB1, CD29 | |
Monoclonal | |
FITC | |
ITGB1 | |
4633401G24Rik; AA409975; AA960159; Beta OL; beta oligodendroglia; CD29; CSAT antigen; ENSMUSG00000051907; Fibronectin receptor subunit beta; FNRB; Glycoprotein IIa; Gm9863; gpIIa; integrin beta 1; integrin beta 1 (fibronectin receptor beta); integrin beta 1A precursor; Integrin beta1; integrin beta-1; integrin beta-1 subunit; integrin beta1D; integrin precursor; integrin subunit beta 1; integrin VLA-4 beta subunit; integrin VLA-4 subunit beta; integrin, beta 1; integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includes MDF2, MSK12); ITBG1D; ITGB1; JG22 antigen; MDF2; MSK12; RGD-receptor; very late activation protein, beta polypeptide; VLA-4 subunit beta; VLAB; VLA-BETA | |
Mouse | |
purified | |
ASR | |
3688 | |
4° C, store in dark | |
Liquid |
Flow Cytometry | |
MEM-101A | |
PBS with BSA and 0.1% sodium azide | |
P05556 | |
ITGB1 | |
Human CD29 | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |