missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ITGA8 Monoclonal Antibody (481709), APC
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA523593
This item is not returnable.
View return policy
Description
ITGA8 Monoclonal Antibody for Flow
Integrin alpha-8/beta-1 functions in the genesis of kidney and probably of other organs by regulating the recruitment of mesenchymal cells into epithelial structures. It recognizes the sequence R-G-D in a wide array of ligands including TNC, FN1, SPP1 TGF.
Specifications
| ITGA8 | |
| Monoclonal | |
| APC | |
| ITGA8 | |
| AI447669; integrin alpha 8; integrin alpha-8; Integrin alpha-8 heavy chain; Integrin alpha-8 light chain; integrin subunit alpha 8; integrin, alpha 8; ITGA8; RGD1564327; zgc:172265 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 8516 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Flow Cytometry | |
| 481709 | |
| Buffered saline solution with ≤ 0.5% BSA and 0.09% sodium azide | |
| P53708 | |
| ITGA8 | |
| E. coli-derived recombinant human Integrin alpha 8 Glu610-Leu800. | |
| 100 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction