Learn More
Invitrogen™ ITGA5/ITGB1 Recombinant Rabbit Monoclonal Antibody (008)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA530937
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human ITGA5 & ITGB1 Heterodimer.
Specifications
| ITGA5/ITGB1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P05556, P08648 | |
| ITGA5, ITGB1 | |
| Recombinant Human ITGA5 & ITGB1 Heterodimer protein. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA | |
| 008 | |
| Unconjugated | |
| ITGA5 | |
| 4633401G24Rik; AA409975; AA960159; Beta OL; beta oligodendroglia; CD29; CD49 antigen-like family member E; Cd49e; ENSMUSG00000051907; fibronectin receptor alpha polypeptide; fibronectin receptor subunit alpha; Fibronectin receptor subunit beta; fibronectin receptor, alpha polypeptide; fibronectin receptor, alpha subunit; FNRA; FNRB; glycoprotein IIa; Gm9863; gpIIa; integrin alpha 5; integrin alpha 5 (fibronectin receptor alpha); integrin alpha-5; Integrin alpha-5 heavy chain; Integrin alpha-5 light chain; Integrin alpha-F; integrin beta 1; integrin beta 1 (fibronectin receptor beta); Integrin beta1; integrin beta-1; integrin subunit alpha 5; integrin subunit beta 1; integrin VLA-4 beta subunit; integrin VLA-4 subunit beta; integrin, alpha 5 (fibronectin receptor, alpha polypeptide); integrin, beta 1; integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includes MDF2, MSK12); Itga5; Itga5_retired; ITGB1; MDF2; MSK12; very late activation protein 5, alpha subunit; very late activation protein, beta polypeptide; VLA-4 subunit beta; VLA5; VLA-5; VLA5A; VLAB; VLA-BETA | |
| Rabbit | |
| Protein A | |
| RUO | |
| 3678, 3688 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.