Learn More
Invitrogen™ ITGA4 Monoclonal Antibody (TA-2)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA49D7
Description
MA49D7 targets CD49d in FACS, IHC, and Neu applications and shows reactivity with Rat and Human samples. The MA49D7 immunogen is rat 150 kD alpha4 chain of VLA-4 (CD49d). MA49D7 detects CD49d which has a predicted molecular weight of approximately 111 kDa. This product has been tested for endotoxins by limulus amoebocyte lysate (LAL) assay and contains an endotoxin concentration of less than or equal to 10 endotoxin units per milligram (EU/mg).
The integrin alpha 4 (also known as CD49d and ITGA4) belongs to the integrin alpha chain family of proteins. Integrins are heterodimeric integral membrane proteins composed of an alpha and beta chains. Alpha4 chain associates with either beta1 or beta7 chain. It has been demonstrated that the putative ligand-binding sites of both integrin alpha4beta1 and alpha4beta7 is located on the alpha4 chain. These ligands included Madcam, VCAM, and fibronectin. Madcam is known as the principal ligand for integrin alpha4beta7. Recently it was also demonstrated that HIV-1 envelope can mimic Madcam by binding to and signaling though integrin alpha4beta7, the gut mucosal homing receptor for peripheral T cells.
Specifications
| ITGA4 | |
| Monoclonal | |
| 1.0 mg/mL | |
| PBS with no preservative | |
| P13612 | |
| ITGA4 | |
| Rat 150 kD alpha4 chain of VLA-4 (CD49d). | |
| 500 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Neutralization | |
| TA-2 | |
| Unconjugated | |
| ITGA4 | |
| 269C wild type; alpha 4 subunit of VLA-4 receptor; antigen CD49D, alpha-4 subunit of VLA-4 receptor; CD49 antigen-like family member D; CD49d; IA4; integrin alpha 4; integrin alpha-4; integrin alpha4B; Integrin alpha-IV; integrin subunit alpha 4; integrin, alpha 4; integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor); Itga4; Itga4B; ITGA-IV; LPAM subunit alpha; lymphocyte Peyer patch adhesion molecules subunit alpha; very late activation protein 4 receptor, alpha 4 subunit; VLA-4 receptor, alpha 4 subunit; VLA-4 subunit alpha | |
| Mouse | |
| Purified | |
| RUO | |
| 311144, 3676 | |
| -20°C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.