missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ITGA10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567829
This item is not returnable.
View return policy
Description
Predicted to react with Mouse samples.
Integrin alpha-10/beta-1 is a receptor for collagen.
Specifications
| ITGA10 | |
| Polyclonal | |
| Unconjugated | |
| ITGA10 | |
| A630048L14; integrin alpha 10; integrin alpha-10; integrin subunit alpha 10; integrin, alpha 10; ITGA10; PRO827; UNQ468/PRO827 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 213119, 8515 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O75578 | |
| ITGA10 | |
| Synthesized peptide derived from human ITGA10. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction