missing translation for 'onlineSavingsMsg'
Learn More

ITFG3 Antibody (OTI3B3), DyLight 594, Novus Biologicals™

Product Code. 18428695 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18428695 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18428695 Supplier Novus Biologicals Supplier No. NBP272253DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ITFG3 Monoclonal antibody specifically detects ITFG3 in Human samples. It is validated for Western Blot, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen ITFG3
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI3B3
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias C16orf9, chromosome 16 open reading frame 9, DKFZP761D0211, FLJ32603, gs19, integrin alpha FG-GAP repeat containing 3, protein ITFG3
Host Species Mouse
Immunogen Full length recombinant protein of human ITFG3 (NP_114428) produced in HEK293T cells.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83986
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.