missing translation for 'onlineSavingsMsg'
Learn More

ITFG3 Antibody (OTI3B3), CoraFluor™ 1, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals (Bio-Techne) NBP2-72253CL1

Product Code. 30150139

  • £870.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ITFG3 Monoclonal antibody specifically detects ITFG3 in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ITFG3
Monoclonal
CoraFluor 1
C16orf9, chromosome 16 open reading frame 9, DKFZP761D0211, FLJ32603, gs19, integrin alpha FG-GAP repeat containing 3, protein ITFG3
Full length human recombinant protein of human ITFG3 (NP_114428) produced in HEK293T cell.
0.1 mL
Primary
Human
Purified
Immunohistochemistry, Western Blot
OTI3B3
PBS
Mouse
Immunogen affinity purified
RUO
83986
Store at 4°C in the dark. Do not freeze.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.